Assay Details
| Gene Symbol: OBP53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001237757.1 | XP_001237758.1 | 2 - 3 | 204 | 83 | |
| XM_316133.3 | XP_316133.3 | 3 - 4 | 219 | 83 | ||
More Information
Set Membership: |
|

OBP53
Genomic Map