Assay Details
| Gene Symbol: MOCOS_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_310528.5 | XP_310528.5 | 1 - 2 | 204 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MOCOS_ANOGA| Gene Symbol: MOCOS_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_310528.5 | XP_310528.5 | 1 - 2 | 204 | 73 | |
Set Membership: |
|