Assay Details
| Gene Symbol: GPRNPY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_310762.5 | XP_310762.5 | 2 - 3 | 859 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPRNPY1| Gene Symbol: GPRNPY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_310762.5 | XP_310762.5 | 2 - 3 | 859 | 69 | |
Set Membership: |
|