Assay Details
| Gene Symbol: MED27_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_312260.4 | XP_312260.3 | 3 - 4 | 790 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED27_ANOGA| Gene Symbol: MED27_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_312260.4 | XP_312260.3 | 3 - 4 | 790 | 61 | |
Set Membership: |
|