Assay Details
| Gene Symbol: SPAST_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_312634.5 | XP_312634.5 | 3 - 4 | 1170 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPAST_ANOGA| Gene Symbol: SPAST_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_312634.5 | XP_312634.5 | 3 - 4 | 1170 | 67 | |
Set Membership: |
|