Assay Details
| Gene Symbol: MED22_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_313084.5 | XP_313084.3 | 1 - 2 | 171 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED22_ANOGA| Gene Symbol: MED22_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_313084.5 | XP_313084.3 | 1 - 2 | 171 | 83 | |
Set Membership: |
|