Assay Details
| Gene Symbol: ANCE9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_313865.5 | XP_313865.3 | 3 - 4 | 2582 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANCE9| Gene Symbol: ANCE9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_313865.5 | XP_313865.3 | 3 - 4 | 2582 | 71 | |
Set Membership: |
|