Assay Details
| Gene Symbol: 1275397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_314636.3 | XP_314636.3 | 3 - 4 | 1050 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

1275397| Gene Symbol: 1275397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_314636.3 | XP_314636.3 | 3 - 4 | 1050 | 102 | |
Set Membership: |
|