Assay Details
| Gene Symbol: GALE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_315024.4 | XP_315024.4 | 4 - 5 | 3779 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GALE3| Gene Symbol: GALE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_315024.4 | XP_315024.4 | 4 - 5 | 3779 | 93 | |
Set Membership: |
|