Assay Details
| Gene Symbol: 1275989 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_315285.4 | XP_315285.3 | 4 - 5 | 1945 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

1275989| Gene Symbol: 1275989 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_315285.4 | XP_315285.3 | 4 - 5 | 1945 | 99 | |
Set Membership: |
|