Assay Details
| Gene Symbol: CPR151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001238068.3 | XP_001238069.3 | 1 - 2 | 293 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPR151| Gene Symbol: CPR151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001238068.3 | XP_001238069.3 | 1 - 2 | 293 | 70 | |
Set Membership: |
|