Assay Details
| Gene Symbol: BOR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332166.1 | NP_001319010.1 | 11 - 12 | 2120 | 84 | |
| NM_001332167.1 | NP_001322448.1 | 11 - 12 | 2280 | 84 | ||
| NM_001332168.1 | NP_001322449.1 | 10 - 11 | 1935 | 84 | ||
| NM_101415.4 | NP_172999.1 | 12 - 13 | 2130 | 84 | ||
| GenBank mRNA | AY069887.1 | - | 11 - 12 | 2045 | 84 | |
More Information
Set Membership: |
|

BOR4
Genomic Map