Assay Details
| Gene Symbol: BOR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332166.1 | NP_001319010.1 | 8 - 9 | 1382 | 144 | |
| NM_001332167.1 | NP_001322448.1 | 9 - 10 | 1393 | 144 | ||
| NM_001332168.1 | NP_001322449.1 | 7 - 8 | 1197 | 144 | ||
| NM_101415.4 | NP_172999.1 | 9 - 10 | 1392 | 144 | ||
| GenBank mRNA | AY069887.1 | - | 8 - 9 | 1307 | 144 | |
More Information
Set Membership: |
|

BOR4
Genomic Map