Assay Details
| Gene Symbol: IAA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101427.5 | NP_173011.1 | 1 - 2 | 382 | 100 | |
| GenBank mRNA | AY150496.1 | - | 1 - 2 | 345 | 100 | |
More Information
Set Membership: |
|

IAA5
Genomic Map