Assay Details
| Gene Symbol: AVP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084073.1 | NP_001077542.1 | 5 - 6 | 1766 | 75 | |
| NM_101437.5 | NP_173021.1 | 5 - 6 | 1830 | 75 | ||
| GenBank mRNA | BT002481.1 | - | 5 - 6 | 1766 | 75 | |
| M81892.1 | - | 5 - 6 | 1796 | 75 | ||
More Information
Set Membership: |
|

AVP1
Genomic Map