Assay Details
| Gene Symbol: TRFL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101440.3 | NP_173024.1 | 1 | 833 | 78 | |
| GenBank mRNA | AK227390.1 | - | 1 | 833 | 78 | |
| AY519536.1 | - | 1 | 720 | 78 | ||
| BT003085.1 | - | 1 | 720 | 78 | ||
More Information
Set Membership: |
|

TRFL5
Genomic Map