Assay Details
| Gene Symbol: AT1G15810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101449.5 | NP_563982.1 | 2 - 3 | 1229 | 81 | |
| GenBank mRNA | AF361614.1 | - | 2 - 3 | 1000 | 81 | |
| AY133600.1 | - | 2 - 3 | 953 | 81 | ||
More Information
Set Membership: |
|

AT1G15810
Genomic Map