Assay Details
| Gene Symbol: AT1G15940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101462.5 | NP_173046.2 | 13 - 14 | 2324 | 123 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G15940| Gene Symbol: AT1G15940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101462.5 | NP_173046.2 | 13 - 14 | 2324 | 123 | |
Set Membership: |
|