Assay Details
| Gene Symbol: CCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084075.1 | NP_001077544.1 | 4 - 5 | 927 | 68 | |
| NM_001332191.1 | NP_001319013.1 | 4 - 5 | 984 | 68 | ||
| NM_001332195.1 | NP_001322098.1 | 3 - 4 | 975 | 68 | ||
| NM_101463.4 | NP_173047.1 | 4 - 5 | 927 | 68 | ||
| GenBank mRNA | AF321114.1 | - | 4 - 5 | 826 | 68 | |
| AF332459.1 | - | 4 - 5 | 826 | 68 | ||
| AK228419.1 | - | 4 - 5 | 925 | 68 | ||
| AY087316.1 | - | 4 - 5 | 928 | 68 | ||
| AY743921.1 | - | 4 - 5 | 826 | 68 | ||
More Information
Set Membership: |
|

CCR1
Genomic Map