Assay Details
| Gene Symbol: ADAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332204.1 | NP_001319016.1 | 2 - 3 | 183 | 72 | |
| NM_001332206.1 | NP_001320849.1 | 1 - 2 | 388 | 72 | ||
| NM_101474.3 | NP_563990.1 | 4 - 5 | 626 | 72 | ||
| NM_202110.2 | NP_973839.1 | 3 - 4 | 849 | 72 | ||
| GenBank mRNA | AY045915.1 | - | 4 - 5 | 430 | 72 | |
| AY133880.1 | - | 4 - 5 | 374 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001332204.1, NM_001332206.1 transcripts.
Set Membership: |
|

ADAP
Genomic Map