Assay Details
| Gene Symbol: ADAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332204.1 | NP_001319016.1 | 5 - 6 | 384 | 83 | |
| NM_001332205.1 | NP_001320850.1 | 5 - 6 | 1050 | 83 | ||
| NM_001332206.1 | NP_001320849.1 | 4 - 5 | 589 | 83 | ||
| NM_001332207.1 | NP_001320852.1 | 3 - 4 | 245 | 83 | ||
| NM_001332208.1 | NP_001320851.1 | 1 - 2 | 156 | 83 | ||
| NM_101474.3 | NP_563990.1 | 7 - 8 | 827 | 83 | ||
| NM_202110.2 | NP_973839.1 | 6 - 7 | 1050 | 83 | ||
| GenBank mRNA | AY045915.1 | - | 7 - 8 | 631 | 83 | |
| AY133880.1 | - | 7 - 8 | 575 | 83 | ||
More Information
Set Membership: |
|

ADAP
Genomic Map