Assay Details
| Gene Symbol: SYP51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035977.1 | NP_001031054.1 | 5 - 6 | 759 | 100 | |
| NM_001332216.1 | NP_001323228.1 | 5 - 6 | 738 | 100 | ||
| NM_101490.4 | NP_563994.1 | 5 - 6 | 785 | 100 | ||
| GenBank mRNA | AF355755.1 | - | 5 - 6 | 695 | 100 | |
| AK317530.1 | - | 5 - 6 | 760 | 100 | ||
| AY074863.1 | - | 5 - 6 | 735 | 100 | ||
| AY085320.1 | - | 5 - 6 | 709 | 100 | ||
| AY097382.1 | - | 4 - 5 | 569 | 100 | ||
More Information
Set Membership: |
|

SYP51
Genomic Map