Assay Details
| Gene Symbol: RDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035979.1 | NP_001031056.1 | 10 - 11 | 1020 | 64 | |
| NM_001198083.1 | NP_001185012.1 | 11 - 12 | 1093 | 64 | ||
| NM_101511.6 | NP_563998.1 | 10 - 11 | 1064 | 64 | ||
| NM_179344.2 | NP_849675.1 | 10 - 11 | 1004 | 64 | ||
| GenBank mRNA | AB032865.1 | - | 10 - 11 | 926 | 64 | |
| AJ010500.1 | - | 10 - 11 | 934 | 64 | ||
| AK229282.1 | - | 10 - 11 | 1023 | 64 | ||
| AY084558.1 | - | 10 - 11 | 1004 | 64 | ||
| BT024876.1 | - | 9 - 10 | 861 | 64 | ||
More Information
Set Membership: |
|

RDH2
Genomic Map