Assay Details
| Gene Symbol: RDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035979.1 | NP_001031056.1 | 11 - 12 | 1089 | 82 | |
| NM_001198083.1 | NP_001185012.1 | 12 - 13 | 1162 | 82 | ||
| NM_001332226.1 | NP_001322506.1 | 10 - 11 | 1134 | 82 | ||
| NM_001332227.1 | NP_001322507.1 | 10 - 11 | 1144 | 82 | ||
| NM_101511.6 | NP_563998.1 | 11 - 12 | 1133 | 82 | ||
| NM_179344.2 | NP_849675.1 | 11 - 12 | 1073 | 82 | ||
| GenBank mRNA | AB032865.1 | - | 11 - 12 | 995 | 82 | |
| AJ010500.1 | - | 11 - 12 | 1003 | 82 | ||
| AK229282.1 | - | 11 - 12 | 1092 | 82 | ||
| AY084558.1 | - | 11 - 12 | 1073 | 82 | ||
| BT024876.1 | - | 10 - 11 | 930 | 82 | ||
More Information
Set Membership: |
|

RDH2
Genomic Map