Assay Details
| Gene Symbol: RDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035979.1 | NP_001031056.1 | 7 - 8 | 819 | 77 | |
| NM_001198083.1 | NP_001185012.1 | 8 - 9 | 892 | 77 | ||
| NM_001332226.1 | NP_001322506.1 | 7 - 8 | 803 | 77 | ||
| NM_001332227.1 | NP_001322507.1 | 7 - 8 | 813 | 77 | ||
| NM_101511.6 | NP_563998.1 | 7 - 8 | 863 | 77 | ||
| NM_179344.2 | NP_849675.1 | 7 - 8 | 803 | 77 | ||
| GenBank mRNA | AB032865.1 | - | 7 - 8 | 725 | 77 | |
| AJ010500.1 | - | 7 - 8 | 733 | 77 | ||
| AK229282.1 | - | 7 - 8 | 822 | 77 | ||
| AY084558.1 | - | 7 - 8 | 803 | 77 | ||
| BT024876.1 | - | 6 - 7 | 660 | 77 | ||
More Information
Set Membership: |
|

RDH2
Genomic Map