Assay Details
Gene Symbol: RDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035979.1 | NP_001031056.1 | 8 - 9 | 874 | 104 | |
NM_001198083.1 | NP_001185012.1 | 9 - 10 | 947 | 104 | ||
NM_001332226.1 | NP_001322506.1 | 8 - 9 | 858 | 104 | ||
NM_001332227.1 | NP_001322507.1 | 8 - 9 | 868 | 104 | ||
NM_101511.6 | NP_563998.1 | 8 - 9 | 918 | 104 | ||
NM_179344.2 | NP_849675.1 | 8 - 9 | 858 | 104 | ||
GenBank mRNA | AB032865.1 | - | 8 - 9 | 780 | 104 | |
AJ010500.1 | - | 8 - 9 | 788 | 104 | ||
AK229282.1 | - | 8 - 9 | 877 | 104 | ||
AY084558.1 | - | 8 - 9 | 858 | 104 | ||
BT024876.1 | - | 7 - 8 | 715 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
