Assay Details
| Gene Symbol: ABA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332230.1 | NP_001322901.1 | 3 - 4 | 357 | 87 | |
| NM_001332231.1 | NP_001322902.1 | 3 - 4 | 357 | 87 | ||
| NM_101519.3 | NP_564001.1 | 3 - 4 | 357 | 87 | ||
| GenBank mRNA | AF325457.1 | - | 3 - 4 | 312 | 87 | |
| AY034895.1 | - | 3 - 4 | 262 | 87 | ||
More Information
Set Membership: |
|

ABA3
Genomic Map