Assay Details
| Gene Symbol: SR45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198085.1 | NP_001185014.1 | 11 - 12 | 1366 | 115 | |
| NM_101523.4 | NP_173107.1 | 11 - 12 | 1322 | 115 | ||
| NM_202115.3 | NP_973844.1 | 11 - 12 | 1301 | 115 | ||
| GenBank mRNA | AF151366.1 | - | 11 - 12 | 1204 | 115 | |
| BT020372.1 | - | 11 - 12 | 1167 | 115 | ||
| BT021095.1 | - | 11 - 12 | 1166 | 115 | ||
More Information
Set Membership: |
|

SR45
Genomic Map