Assay Details
| Gene Symbol: SR45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198085.1 | NP_001185014.1 | 2 - 3 | 435 | 104 | |
| NM_001332238.1 | NP_001321127.1 | 2 - 3 | 424 | 104 | ||
| NM_001332239.1 | NP_001321128.1 | 2 - 3 | 456 | 104 | ||
| NM_001332240.1 | NP_001321125.1 | 2 - 3 | 439 | 104 | ||
| NM_001332241.1 | NP_001321126.1 | 2 - 3 | 439 | 104 | ||
| NM_101523.4 | NP_173107.1 | 2 - 3 | 424 | 104 | ||
| NM_202115.3 | NP_973844.1 | 2 - 3 | 424 | 104 | ||
| GenBank mRNA | AF151366.1 | - | 2 - 3 | 306 | 104 | |
| BT020372.1 | - | 2 - 3 | 269 | 104 | ||
| BT021095.1 | - | 2 - 3 | 289 | 104 | ||
More Information
Set Membership: |
|

SR45
Genomic Map