Assay Details
| Gene Symbol: AT1G16680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101528.4 | NP_173112.2 | 4 - 5 | 1806 | 119 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G16680| Gene Symbol: AT1G16680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101528.4 | NP_173112.2 | 4 - 5 | 1806 | 119 | |
Set Membership: |
|