Assay Details
| Gene Symbol: AT1G16680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101528.4 | NP_173112.2 | 5 - 6 | 1912 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G16680| Gene Symbol: AT1G16680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101528.4 | NP_173112.2 | 5 - 6 | 1912 | 60 | |
Set Membership: |
|