Assay Details
| Gene Symbol: HAC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198086.1 | NP_001185015.1 | 14 - 15 | 4537 | 78 | |
| NM_001332245.1 | NP_001321740.1 | 14 - 15 | 4777 | 78 | ||
| NM_001332246.1 | NP_001321737.1 | 14 - 15 | 4987 | 78 | ||
| NM_001332247.1 | NP_001321739.1 | 14 - 15 | 4870 | 78 | ||
| NM_001332248.1 | NP_001321738.1 | 15 - 16 | 4714 | 78 | ||
| NM_001332249.1 | NP_001321742.1 | 14 - 15 | 4475 | 78 | ||
| NM_001332250.1 | NP_001321741.1 | 14 - 15 | 4543 | 78 | ||
| NM_001332251.1 | NP_001321736.1 | 14 - 15 | 4823 | 78 | ||
| NM_001332252.1 | NP_001321735.1 | 14 - 15 | 4804 | 78 | ||
| NM_101532.3 | NP_173115.1 | 13 - 14 | 4570 | 78 | ||
More Information
Set Membership: |
|

HAC12
Genomic Map