Assay Details
| Gene Symbol: EBS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101551.4 | NP_173134.2 | 3 - 4 | 654 | 80 | |
| GenBank mRNA | AY140061.1 | - | 3 - 4 | 654 | 80 | |
| BT010395.1 | - | 3 - 4 | 524 | 80 | ||
| KJ139003.1 | - | 3 - 4 | 524 | 80 | ||
More Information
Set Membership: |
|

EBS3
Genomic Map