Assay Details
| Gene Symbol: KU70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101558.4 | NP_564012.1 | 12 - 13 | 1298 | 71 | |
| GenBank mRNA | AF283759.1 | - | 12 - 13 | 1261 | 71 | |
| AK221642.1 | - | 12 - 13 | 1286 | 71 | ||
More Information
Set Membership: |
|

KU70
Genomic Map