Assay Details
| Gene Symbol: KU70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101558.4 | NP_564012.1 | 13 - 14 | 1359 | 61 | |
| GenBank mRNA | AF283759.1 | - | 13 - 14 | 1322 | 61 | |
| AK221642.1 | - | 13 - 14 | 1347 | 61 | ||
More Information
Set Membership: |
|

KU70
Genomic Map