Assay Details
| Gene Symbol: KU70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101558.4 | NP_564012.1 | 7 - 8 | 767 | 105 | |
| GenBank mRNA | AF283759.1 | - | 7 - 8 | 730 | 105 | |
| AK221642.1 | - | 7 - 8 | 755 | 105 | ||
More Information
Set Membership: |
|

KU70
Genomic Map