Assay Details
| Gene Symbol: TPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332273.1 | NP_001321461.1 | 10 - 11 | 1533 | 130 | |
| NM_101559.2 | NP_173142.2 | 10 - 11 | 1533 | 130 | ||
More Information
Set Membership: |
|

TPS2
Genomic Map