Assay Details
| Gene Symbol: UBP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198090.2 | NP_001185019.1 | 3 - 4 | 1297 | 95 | |
| NM_001332280.1 | NP_001322494.1 | 3 - 4 | 1435 | 95 | ||
| NM_101571.2 | NP_564014.1 | 3 - 4 | 1252 | 95 | ||
More Information
Set Membership: |
|

UBP15
Genomic Map