Assay Details
| Gene Symbol: UBP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198090.2 | NP_001185019.1 | 8 - 9 | 1883 | 69 | |
| NM_001332280.1 | NP_001322494.1 | 8 - 9 | 2009 | 69 | ||
| NM_101571.2 | NP_564014.1 | 8 - 9 | 1826 | 69 | ||
More Information
Set Membership: |
|

UBP15
Genomic Map