Assay Details
| Gene Symbol: ILP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101582.3 | NP_173164.1 | 3 - 4 | 562 | 77 | |
| GenBank mRNA | AK229089.1 | - | 3 - 4 | 547 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ILP1| Gene Symbol: ILP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101582.3 | NP_173164.1 | 3 - 4 | 562 | 77 | |
| GenBank mRNA | AK229089.1 | - | 3 - 4 | 547 | 77 | |
Set Membership: |
|