Assay Details
| Gene Symbol: ILP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101582.3 | NP_173164.1 | 4 - 5 | 900 | 78 | |
| GenBank mRNA | AK229089.1 | - | 4 - 5 | 885 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ILP1| Gene Symbol: ILP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101582.3 | NP_173164.1 | 4 - 5 | 900 | 78 | |
| GenBank mRNA | AK229089.1 | - | 4 - 5 | 885 | 78 | |
Set Membership: |
|