Assay Details
| Gene Symbol: ILP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101582.3 | NP_173164.1 | 5 - 6 | 2668 | 131 | |
| GenBank mRNA | AK229089.1 | - | 5 - 6 | 2653 | 131 | |
More Information
Set Membership: |
|

ILP1
Genomic Map