Assay Details
| Gene Symbol: EER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101605.5 | NP_564023.1 | 4 - 5 | 1962 | 89 | |
| NM_179349.1 | NP_849680.1 | 4 - 5 | 1799 | 89 | ||
| GenBank mRNA | AY056144.1 | - | 4 - 5 | 1799 | 89 | |
| AY091423.1 | - | 4 - 5 | 1691 | 89 | ||
| AY463614.1 | - | 4 - 5 | 1691 | 89 | ||
More Information
Set Membership: |
|

EER4
Genomic Map