Assay Details
| Gene Symbol: TRFL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198094.1 | NP_001185023.1 | 5 - 6 | 600 | 129 | |
| NM_101608.5 | NP_564025.2 | 4 - 5 | 589 | 129 | ||
| GenBank mRNA | AY080665.1 | - | 4 - 5 | 540 | 129 | |
| AY122966.1 | - | 3 - 4 | 351 | 129 | ||
| AY519537.1 | - | 3 - 4 | 351 | 129 | ||
More Information
Set Membership: |
|

TRFL3
Genomic Map