Assay Details
| Gene Symbol: TRFL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198094.1 | NP_001185023.1 | 9 - 10 | 1912 | 82 | |
| NM_001332306.1 | NP_001321911.1 | 8 - 9 | 1838 | 82 | ||
| NM_101608.5 | NP_564025.2 | 8 - 9 | 1901 | 82 | ||
| GenBank mRNA | AY080665.1 | - | 8 - 9 | 1852 | 82 | |
| AY122966.1 | - | 7 - 8 | 1663 | 82 | ||
| AY519537.1 | - | 7 - 8 | 1663 | 82 | ||
More Information
Set Membership: |
|

TRFL3
Genomic Map