Assay Details
| Gene Symbol: CSTF77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101639.3 | NP_173218.2 | 1 - 2 | 402 | 52 | |
| GenBank mRNA | BT002320.1 | - | 1 - 2 | 56 | 52 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSTF77| Gene Symbol: CSTF77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101639.3 | NP_173218.2 | 1 - 2 | 402 | 52 | |
| GenBank mRNA | BT002320.1 | - | 1 - 2 | 56 | 52 | |
Set Membership: |
|