Assay Details
| Gene Symbol: PAPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101661.5 | NP_173240.2 | 10 - 11 | 2329 | 91 | |
| NM_202126.2 | NP_973855.1 | 8 - 9 | 2364 | 91 | ||
| GenBank mRNA | AK227446.1 | - | 10 - 11 | 2329 | 91 | |
| AY323906.1 | - | 10 - 11 | 2039 | 91 | ||
| BT005990.1 | - | 10 - 11 | 2039 | 91 | ||
More Information
Set Membership: |
|

PAPS1
Genomic Map