Assay Details
| Gene Symbol: CDKD1;3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101666.3 | NP_173244.1 | 2 - 3 | 626 | 90 | |
| GenBank mRNA | AB047274.1 | - | 2 - 3 | 509 | 90 | |
| AY099677.1 | - | 2 - 3 | 598 | 90 | ||
| AY128857.1 | - | 2 - 3 | 397 | 90 | ||
More Information
Set Membership: |
|

CDKD1;3
Genomic Map