Assay Details
| Gene Symbol: E12A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101672.4 | NP_173250.1 | 1 - 2 | 282 | 64 | |
| GenBank mRNA | AF147721.1 | - | 1 - 2 | 250 | 64 | |
| AF332406.1 | - | 1 - 2 | 198 | 64 | ||
More Information
Set Membership: |
|

E12A11
Genomic Map