Assay Details
| Gene Symbol: E12A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101672.4 | NP_173250.1 | 2 - 3 | 338 | 59 | |
| GenBank mRNA | AF147721.1 | - | 2 - 3 | 306 | 59 | |
| AF332406.1 | - | 2 - 3 | 254 | 59 | ||
More Information
Set Membership: |
|

E12A11
Genomic Map